Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP4 Mouse anti-Human, Mouse, Rat, Clone: S164-6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527674

Catalog No. PIMA527674


Add to cart

Description

Description

This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cei-Lenz syndrome.
Specifications

Specifications

LRP4
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
O75096, Q8VI56, Q9QYP1
Corin, KIAA0816, LDLR, LDLR dan, Low density lipoprotein receptor related protein 4, LRP-4, LRP10, MEGF7, Multiple epidermal growth factor like domains 7
Mouse
IgG1
100 μg
-20°C
Primary
228357, 4038, 83469
Immunocytochemistry, Immunofluorescence, Western Blot
S164-6
Unconjugated
LRP4
Liquid
LRP4
Fusion protein amino acids 1747-1905 (cytoplasmic C-terminus) of mouse LRP4
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit