Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lrp4 Mouse anti-Human, Mouse, Rat, FITC, Clone:S164-6, Abnova™

Mouse monoclonal antibody raised against partial recombinant mouse Lrp4.

Manufacturer:  Abnova Corporation MAB16815

Catalog No. 89-333-439


Add to cart

Specifications

Specifications

Lrp4
S164-6
low density lipoprotein receptor-related protein 4
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Liquid
Mouse
IgG1
Store at -20°C.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
FITC
Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Lrp4
6430526J12Rik, D230026E03, Megf7, mdig
Recombinant protein corresponding to amino acids 1747-1905 at C-terminus of mouse Lrp4.
100μg
Antibody
228357
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit