Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRP4 Polyclonal Antibody
LRP4 Polyclonal Antibody
Supplier: Thermo Scientific PA547795
Description
LRP4 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
LRP4 | |
Polyclonal | |
Unconjugated | |
LRP4 | |
6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2 | |
Sheep | |
Antigen Affinity Chromatography | |
RUO | |
4038, 83469 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O75096, Q9QYP1 | |
LRP4 | |
E. coli-derived recombinant human LRP-4 Gly18-Ser344. | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction