Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRPAP Mouse anti-Human, Alexa Fluor™ 594, Clone: 8G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242682AF594

Catalog No. NP242682A94


Add to cart

Description

Description

LRPAP Monoclonal antibody specifically detects LRPAP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

LRPAP
8G8
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Purified, Conjugated
LRPAP1
Recombinant human LRPAP1 (35-357aa) purified from E. coli
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP
Mouse
IgG1 Kappa
100 μL
Primary
4043.0
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit