Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRPAP Antibody (8G8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242682H
Description
LRPAP Monoclonal specifically detects LRPAP in Human samples. It is validated for Western Blot, ELISA.Specifications
LRPAP | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LRPAP1 | |
Recombinant human LRPAP1 (35-357aa) purified from E. coli | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
8G8 | |
Western Blot, ELISA | |
A2MRAPMRAP, A2RAP, alpha-2-macroglobulin receptor-associated protein, alpha-2-macroglobulin receptor-associated protein 1, Alpha-2-MRAP, HBP44, lipoprotein receptor associated protein, low density lipoprotein receptor-related protein associated protein 1, Low density lipoprotein receptor-related protein-associated protein 1, low density lipoprotein-related protein-associated protein 1(alpha-2-macroglobulin receptor-associated protein 1), MGC138272, RAP | |
Mouse | |
Protein A purified | |
RUO | |
4043 | |
Store at 4C in the dark. | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction