Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRPAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 246621AP150UL
Description
Interacts with LRP1/alpha-2-macroglobulin receptor and glycoprotein 330.Specifications
LRPAP1 | |
Polyclonal | |
Unconjugated | |
LRPAP1 | |
A2MRAP, A2RAP, Alpha 2 MRAP, HBP44, LRPAP1, MRAP, RAP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4043 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P30533 | |
LRPAP1 | |
LRPAP1 Fusion Protein Ag20304 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction