Learn More
Aviva Systems Biology Corp LRRC15 Antibody - N-terminal region (ARP50292_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50292_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC15. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRRC15. Alternate names: LIBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60310