Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550442
Description
The antibody detects endogenous levels of total LRRC15 protein.
LRRC15 Polyclonal antibody specifically detects LRRC15 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
LRRC15 | |
Polyclonal | |
Unconjugated | |
LRRC15 | |
5430427N11Rik; hLib; leucine rich repeat containing 15; Leucine-rich repeat protein induced by amyloid-beta; leucine-rich repeat protein induced by beta amyloid; leucine-rich repeat protein induced by beta-amyloid; leucine-rich repeat protein induced by beta-amyloid homolog; Leucine-rich repeat-containing protein 15; LIB; Lrrc15; rLib | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131578 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8TF66 | |
LRRC15 | |
Fusion protein corresponding to a region derived from internal residues of human leucine rich repeat containing 15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction