Learn More
LRRC17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209181AP
Catalog No. 50-173-3154
Add to cart
Description
Description
Leucine-rich repeat-containing protein 17 (LRRC17) is an LRR protein predomitly expressed by osteoblasts. LRRC17 acts as a negative regulator of receptor activator of NF-kappaB ligand (RANKL)-induced osteoclast differentiation. It has been reported that low plasma level of LRRC17 is an independent risk factor for osteoporotic fractures.
Specifications
Specifications
LRRC17
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N6Y2, Q9CXD9
LRRC17, P37NB
Rabbit
IgG
150 μL
-20°C
Primary
10234, 74511
Western Blot
Unconjugated
LRRC17
Liquid
Lrrc17
LRRC17 Fusion Protein Ag15117
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse