Learn More
Abnova™ LRRC20 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00055222T01
Catalog No. 89-951-110
Add to cart
Description
Description
Specifications
Specifications
293T
NM_207119
pCMV-LRRC20 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LRRC20 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 20.5kDa
Transient overexpression cell lysate was tested with anti-LRRC20 antibody (Mfr. No. H00055222-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human