Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563449
Description
Immunogen sequence: LKADGNRLRS AQMNELPYLQ IASFAYNQIT DTEGISHPRL ETLNLKGNSI HMVTGLDPEK LISLHTVELR GNQLESTLGI NLPKLKNLYL A Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

Specifications
LRRC23 | |
Polyclonal | |
Unconjugated | |
LRRC23 | |
4921537K05Rik; B7; leucine rich protein, B7; leucine rich repeat containing 23; leucine-rich protein B7; Leucine-rich repeat-containing protein 23; LRPB7; LRRC23; neuron-specific enolase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53EV4 | |
LRRC23 | |
Recombinant protein corresponding to Human LRRC23. Recombinant protein control fragment (Product #RP-103301). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction