Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRC26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563285
Description
Immunogen sequence: PEVCTCVPGG LASCSALSLP AVPPGLSLRL RALLLDHNRV RA Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 67%.
The LRRC26 protein was initially identified as a cytokeratin-associated protein whose expression was elevated in multiple cancer cell lines and cancer specimens as well in normal prostate and salivary gland tissues. LRRC26 was later found to negatively regulate the NF-kappaB activation and suppresses tumor growth and metastasis. LRRC26 can also act to selectively alter the efficacy of large conductance calcium-activated K (BK) channels.
Specifications
LRRC26 | |
Polyclonal | |
Unconjugated | |
LRRC26 | |
bA350O14.10; BK channel auxiliary gamma subunit LRRC26; BK channel auxilliary gamma subunit LRRC26; CAPC; cytokeratin associated protein; cytokeratin-associated protein in cancer; leucine rich repeat containing 26; leucine-rich repeat-containing protein 26; Lrrc26; RGD1308398 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
389816 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2I0M4 | |
LRRC26 | |
Recombinant protein corresponding to Human LRRC26. Recombinant protein control fragment (Product #RP-100074). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction