Learn More
Aviva Systems Biology Corp LRRC28 Antibody - C-terminal region (ARP52691_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52691_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LRRC28. Alternate names: FLJ34269, FLJ45242, MGC24976ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60805