Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC32/GARP Antibody (855151), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB6055H
Description
LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| LRRC32/GARP | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 855151 | |
| Flow Cytometry, CyTOF-ready | |
| D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
| recombinant human LRRC32/GARP, Accession # Q14392 | |
| RUO | |
| 2615 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction