Learn More
LRRC32/GARP Mouse anti-Human, Alexa Fluor™ 532, Clone: GARP5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237710AF532
Catalog No. NP237710A32
Add to cart
Description
Description
LRRC32/GARP Monoclonal antibody specifically detects LRRC32/GARP in Human samples. It is validated for Western Blot.
Specifications
Specifications
LRRC32/GARP
GARP5
Western Blot
Purified, Conjugated
LRRC32
Purified human sGARP protein.
Protein A purified
Store at 4°C in the dark.
Monoclonal
Antibody recognizes GARP/ LRRC32, an approximately 80 kDa glycoprotein expressed e.g. on the surface of megakaryocytes, platelets and activated Treg cells.
Western Blot
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Mouse
IgG1
0.1 mL
Primary
2615.0
Human