Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC38 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548304

Catalog No. PA548304

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Auxiliary protein of the large-conductance, voltage and calcium-activated potassium channel (BK alpha). Modulates gating properties by producing a marked shift in the BK channel ft.s voltage dependence of activation in the hyperpolarizing direction, and in the absence of calcium.
Specifications

Specifications

LRRC38
Polyclonal
PBS with 0.09% sodium azide
Q5VT99
Leucine-rich repeat-containing protein 38, BK channel auxiliary gamma subunit LRRC38, LRRC38
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
126755
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LRRC38
Liquid
LRRC38
KLH conjugated synthetic peptide between 188-216 amino acids from the C-terminal region of human LRRC38.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit