Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC39 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12312R

Description
Component of the sarcomeric M-band which plays a role in myocyte response to biomechanical stress. May regulate expression of other M-band proteins via an SRF-dependent pathway. Important for normal contractile function in heart.Specifications
LRRC39 | |
Polyclonal | |
Unconjugated | |
LRRC39 | |
densin hlg; leucine rich repeat containing 39; Leucine-rich repeat-containing protein 39; LRRC39; Myomasp; Myosin-interacting M-band-associated stress-responsive protein; UNQ6500/PRO21368 | |
Rabbit | |
Protein A | |
RUO | |
127495 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96DD0 | |
LRRC39 | |
KLH conjugated synthetic peptide derived from human LRRC39. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction