Learn More
Abnova™ LRRC4C 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00057689T01
Catalog No. 89-951-136
Add to cart
Description
Description
Specifications
Specifications
293T
NM_020929
pCMV-LRRC4C full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LRRC4C 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 71.9kDa
Transient overexpression cell lysate was tested with anti-LRRC4C antibody (Mfr. No. H00057689-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human