Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC50 Mouse anti-Human, Clone: OTI3E2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504577

Catalog No. 50-167-5231


Add to cart

Description

Description

The protein encoded by this gene is cilium-specific and is required for the stability of the ciliary architecture. It is involved in the regulation of microtubule-based cilia and actin-based brush border microvilli. Mutations in this gene are associated with primary ciliary dyskinesia-13.
Specifications

Specifications

LRRC50
Monoclonal
1.25 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8NEP3
CILD13, LRRC50, ODA7, swt
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
123872
Western Blot
OTI3E2
Unconjugated
DNAAF1
Liquid
DNAAF1
Full length human recombit protein of human LRRC50 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit