Learn More
Aviva Systems Biology Corp LRRC59 Antibody - middle region (ARP49323_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRC59. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human LRRC59. Alternate names: FLJ21675, PRO1855ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60063