Learn More
Abnova™ LRRC6 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00023639T01
Catalog No. 89-951-147
Add to cart
Description
Description
Specifications
Specifications
293T
NM_012472
pCMV-LRRC6 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LRRC6 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 54.3kDa
Transient overexpression cell lysate was tested with anti-LRRC6 antibody (Mfr. No. H00023639-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human