Learn More
LRRC6 Mouse anti-Human, Clone: OTI7E9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806109
Catalog No. 50-166-7698
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
LRRC6 may be involved in spermatocytogenesis or prophase of meiosis.
Specifications
Specifications
LRRC6
Monoclonal
Unconjugated
LRRC6
Lyophilized
LRRC6
Full length human recombit protein of human LRRC6 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI7E9
PBS with 8% trehalose and no preservative; pH 7.3
Q86X45
CILD19, LRTP, TSLRP
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
23639