Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548269

Catalog No. PA548269

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with monkey based on sequence homology.

LRRC6 may be involved in spermatocytogenesis or prophase of meiosis.
Specifications

Specifications

LRRC6
Polyclonal
PBS with 0.09% sodium azide
Q86X45
Protein tilB homolog, Leucine-rich repeat-containing protein 6, Leucine-rich testis-specific protein, Testis-specific leucine-rich repeat protein, LRRC6, LRTP, TSLRP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23639
Flow Cytometry, Western Blot
Unconjugated
LRRC6
Liquid
LRRC6
KLH conjugated synthetic peptide between 179-207 amino acids from the Central region of human LRRC6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit