Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC63 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571596
Description
The function of this protein remains unknown.Specifications
LRRC63 | |
Polyclonal | |
Unconjugated | |
LRRC63 | |
Leucine-rich repeat-containing protein 63; LRRC63 | |
Rabbit | |
Protein A | |
RUO | |
220416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.89 mg/mL | |
PBS with 0.09% sodium azide | |
Q05C16 | |
LRRC63 | |
KLH conjugated synthetic peptide between 484-513 amino acids from the C-terminal region of human LRRC63 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title