Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC7 Polyclonal Antibody
LRRC7 Polyclonal Antibody
Supplier: Thermo Scientific PA588994
Description
LRRC7 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
LRRC7 | |
Polyclonal | |
Unconjugated | |
LRRC7 | |
AK122498; densin; densin 13T-16; densin 13T-17; densin 13T-23; densin 13T-29; densin 13T-41; densin 13T-43; densin 13T-57; densin 13T-57C; densin 13T-72; densin 13T-89; densin 180; densin-180; Kiaa1365; Lap1; leucine rich repeat containing 7; leucine-rich repeat-containing protein 7; LOW QUALITY PROTEIN: leucine-rich repeat-containing protein 7; Lrrc7; mKIAA1365; Protein LAP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117284, 242274, 57554 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.26 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P70587, Q80TE7, Q96NW7 | |
LRRC7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 730-890 of human LRRC7 (NP_0658451). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction