Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC70 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549118

Catalog No. PA549118

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Renders cells highly sensitive to the activation by cytokines and lipopolysaccharide (LPS).
Specifications

Specifications

LRRC70
Polyclonal
PBS with 0.09% sodium azide
Q7Z2Q7
Leucine-rich repeat-containing protein 70, Synleurin, LRRC70
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100130733
Western Blot
Unconjugated
LRRC70
Liquid
LRRC70
KLH conjugated synthetic peptide between 542-570 amino acids from the C-terminal region of human LRRC70.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit