Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC8A Rabbit anti-Human, Mouse, Alexa Fluor™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP232158AF350

Catalog No. NP23218A30


Add to cart

Description

Description

LRRC8A Polyclonal antibody specifically detects LRRC8A in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

LRRC8A
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
AGM5, FLJ10337, FLJ41617, KIAA1437leucine rich repeat containing 8, leucine rich repeat containing 8 family, member A, leucine-rich repeat-containing protein 8A, LRRC8
Rabbit
IgG
100 μL
Primary
56262.0
Western Blot
Western Blot
Affinity Purified, Conjugated
LRRC8A
The immunogen this antibody was made to, maps to a region between residue 760 to 810 of human Leucine Rich Repeat Containing 8 Family, Member A using the numbering given in entry NP_001120716.1 (GeneID 56262).
Immunogen affinity purified
Store at 4°C in the dark.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit