Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

LRRC8A Rabbit anti-Human, Mouse, DyLight 755, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232158IR

Catalog No. NBP232158IR



Description

Description

LRRC8A Polyclonal antibody specifically detects LRRC8A in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

LRRC8A
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
AGM5, FLJ10337, FLJ41617, KIAA1437leucine rich repeat containing 8, leucine rich repeat containing 8 family, member A, leucine-rich repeat-containing protein 8A, LRRC8
Rabbit
IgG
100 ul
Store at 4C in the dark.
Polyclonal
Human, Mouse
Western Blot
Western Blot
Affinity Purified
LRRC8A
The immunogen this antibody was made to, maps to a region between residue 760 to 810 of human Leucine Rich Repeat Containing 8 Family, Member A using the numbering given in entry NP_001120716.1 (GeneID 56262).
Immunogen affinity purified
RUO
Primary
56262
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only