Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC8E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$461.00
Specifications
Antigen | LRRC8E |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP160062
|
Novus Biologicals
NBP160062 |
100 μL |
Each of 1 for $461.00
|
|
|||||
Description
LRRC8E Polyclonal specifically detects LRRC8E in Human samples. It is validated for Western Blot.Specifications
LRRC8E | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
80131 | |
Synthetic peptides corresponding to LRRC8E(leucine rich repeat containing 8 family, member E) The peptide sequence was selected from the middle region of LRRC8E. Peptide sequence LRELKQLKVLSLRSNAGKVPASVTDVAGHLQRLSLHNDGARLVALNSLKK. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
FLJ23420, leucine rich repeat containing 8 family, member E, leucine-rich repeat-containing protein 8E | |
LRRC8E | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title