Learn More
Aviva Systems Biology Corp LRRN3 Antibody - N-terminal region (ARP47422_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LRRN3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LRRN3. Alternate names: FIGLER5, FLJ11129, NLRR-3, NLRR3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59547