Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRRN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113430
Description
LRRN3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Specifications
LRRN3 | |
Polyclonal | |
Unconjugated | |
LRRN3 | |
fibronectin type III, immunoglobulin and leucine rich repeat domains 5; FIGLER5; leucine rich repeat neuronal 3; leucine rich repeat protein 3, neuronal; leucine-rich repeat neuronal protein 3; leucine-rich repeat protein, neuronal 3; Lrrn3; Nbla10363; Neuronal leucine-rich repeat protein 3; NLRR3; NLRR-3; UNQ194/PRO220 | |
Rabbit | |
Protein G | |
RUO | |
54674 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.196 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9H3W5 | |
LRRN3 | |
Recombinant Human Leucine-rich repeat neuronal protein 3 protein (201-500AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction