Learn More
LRRTM1 Mouse anti-Human, Clone: OTI5H9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805611
Catalog No. 50-166-7503
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM1 is a maternally suppressed gene that is associated paternally with handedness and schizophrenia.
Specifications
Specifications
LRRTM1
Monoclonal
Unconjugated
LRRTM1
Lyophilized
LRRTM1
Human recombit protein fragment corresponding to amino acids 182-422 of human LRRTM1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI5H9
PBS with 8% trehalose and no preservative; pH 7.3
Q86UE6
leucine rich repeat transmembrane neuronal 1
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
347730