Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRTM2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen LRRTM2
Dilution Immunohistochemistry -1:50-1:200, Immunohistochemistry-Paraffin -1:50-1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121446
View Documents
Novus Biologicals
NBP311369200UL
200 μg
Each for $320.71
Add to cart
 
NB121447
View Documents
Novus Biologicals
NBP31136950UL
50 μg
Each for $138.37
Add to cart
 
Description

Description

LRRTM2 Polyclonal antibody specifically detects LRRTM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

LRRTM2
Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
PBS, pH7.0
26045
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry -1:50-1:200, Immunohistochemistry-Paraffin -1:50-1:200
Unconjugated
Rabbit
Human
leucine-rich repeat transmembrane neuronal protein 2, leucine rich repeat transmembrane neuronal 2
E. coli-derived Human LRRTM2 fragment
Polyclonal
Antigen and protein A Affinity-purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit