Learn More
LRSAM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 246661AP
Catalog No. 50-173-4482
Add to cart
Description
Description
LRSAM1 is a multifunctional RING finger protein that selectively regulates cell adhesion molecules, has ubiquitin ligase activity, and plays a role in receptor endocytosis and viral budding.
Specifications
Specifications
LRSAM1
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6UWE0, Q80ZI6
hTAL, LRSAM1, RIFLE, TAL, Tsg101 associated ligase
Rabbit
IgG
150 μL
-20°C
Primary
227738, 90678
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LRSAM1
Liquid
LRSAM1
LRSAM1 Fusion Protein Ag20415
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse