Learn More
LRTM1 Rabbit anti-Human, Alexa Fluor™ 350, Clone: 2381D, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB10046U100UG
Catalog No. FB10046U100
Add to cart
This item is not returnable. View return policy
Description
Description
LRTM1 Monoclonal antibody specifically detects LRTM1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
LRTM1
2381D
Flow Cytometry 0.25-1 ug/10^6 cells
Q9HBL6
HT017, leucine-rich repeat and transmembrane domain-containing protein 1, leucine-rich repeats and transmembrane domains 1
Synthetic peptide containing human LRTM1, Accession No. Q9HBL6
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human LRTM1 in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 350
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Rabbit
IgG
100 μg
Primary
57408.0
Human