Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LRTM1 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB10046N100UG
This item is not returnable.
View return policy
Description
LRTM1 Monoclonal specifically detects LRTM1 in Human samples. It is validated for Flow Cytometry.Specifications
LRTM1 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide | |
HT017, leucine-rich repeat and transmembrane domain-containing protein 1, leucine-rich repeats and transmembrane domains 1 | |
Synthetic peptide containing human LRTM1, Accession No. Q9HBL6 | |
Primary | |
Detects human LRTM1 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Flow Cytometry | |
2381D | |
Flow Cytometry 0.25-1 μg/106 cells | |
Q9HBL6 | |
Rabbit | |
100 μg | |
57408 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction