Learn More
Invitrogen™ LRTOMT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114402
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human MCF-7 whole cell, human Hela whole cell, human Caco-2 whole cell, human K562 whole cell, human U20S whole cell, human THP-1 whole cell, rat brain tissue, rat ovarian tissue, rat heart tissue, rat lung tissue, mouse brain tissue, mouse lung tissue. IHC: human placenta tissue, human Lung cancer tissue, mouse brain tissue, mouse brain tissue, rat brain tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Catalyzes the O-methylation, and thereby the inactivation, of catecholamine neurotransmitters and catechol hormones. Required for auditory function.
Specifications
LRTOMT | |
Polyclonal | |
Unconjugated | |
LRTOMT | |
1700008D07Rik; Catechol O-methyltransferase 2; catechol O-methyltransferase 2 {ECO:0000250; CFAP111; Comt2; DFNB63; leucine rich repeat containing 51; leucine rich transmembrane and 0-methyltransferase domain containing; leucine rich transmembrane and O-methyltransferase domain containing; leucine-rich repeat-containing protein 51; leucine-rich repeat-containing protein 51 {ECO:0000312; leucine-rich repeat-containing protein 51; transmembrane O-methyltransferase; LRRC51; lrrc51 {ECO:0000312; Lrtomt; Lrtomt1; O-methyltransferase domain containing; PP7517; Protein LRTOMT1; protein LRTOMT2; RGD:1561509}; RGD:1565856}; RGD1561509; RGD1565856; Tomt; tomt {ECO:0000312; Transmembrane O-methyltransferase; transmembrane O-methyltransferase {ECO:0000312; Transmembrane O-methyltransferase homolog; UniProtKB:A1Y9I9} | |
Rabbit | |
Affinity chromatography | |
RUO | |
220074, 293156, 308868, 69358 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
B6CZ61, B6CZ62, Q8WZ04, Q9DAK8 | |
Lrrc51, LRTOMT | |
A synthetic peptide corresponding to a sequence of human LRTOMT (RLLTVERDPRTAAVAEKLIRLAGFDEHMVEL). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.