Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRTOMT Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549421

Catalog No. PA549421

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalyzes the O-methylation, and thereby the inactivation, of catecholamine neurotransmitters and catechol hormones. Required for auditory function.
Specifications

Specifications

LRTOMT
Polyclonal
PBS with 0.09% sodium azide
Q8WZ04
Transmembrane O-methyltransferase, Catechol O-methyltransferase 2, Protein LRTOMT2, LRTOMT, COMT2, TOMT
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
220074
Western Blot
Unconjugated
LRTOMT
Liquid
LRTOMT
KLH conjugated synthetic peptide between 2-36 amino acids from the N-terminal region of human LRTOMT.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit