Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRWD1 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571585
Description
Required for G1/S transition. Recruits and stabilizes the origin recognition complex (ORC) onto chromatin during G1 to establish pre-replication complex (preRC) and to heterochromatic sites in post-replicated cells. Binds a combination of DNA and histone methylation repressive marks on heterochromatin. Binds histone H3 and H4 trimethylation marks H3K9me3, H3K27me3 and H4K20me3 in a cooperative manner with DNA methylation. Required for silencing of major satellite repeats. May be important ORC2, ORC3 and ORC4 stability.
Specifications
LRWD1 | |
Polyclonal | |
Unconjugated | |
Lrwd1 | |
CENP-33; ORCA; ORC-associated protein; centromere protein 33; leucine-rich repeat and WD repeat-containing protein 1; origin recognition complex associated; origin recognition complex-associated protein | |
Rabbit | |
Protein A | |
RUO | |
71735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Q8BUI3 | |
Lrwd1 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human LRWD1 | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction