Learn More
Polyscience LS CHILLER, 1160W, 120V/60HZ DFS Item
Manufacturer:  Polyscience LS51M11A110C
LS Benchtop Chiller, -20 to +40C, 1160 W, 14.8 l/min, 9.0 psi, 120V/60Hz, Recirculating Chiller, Chiller, Laser, Rotary Evaporator, Extraction
Catalog No. 50-113-0657