Learn More
Teledyne SSI LS Class Pump
The reliable LS Class consists of single-headed, positive displacement piston pumps incorporating advanced cam designs for very low pulsation. 
$4310.00 - $5230.00
Specifications
For Use With (Application) Dispensing, Metering, Semi-Prep, Separation
View More Specs
Products 9
Catalog Number Mfr. No. Flow Rate Pressure Material Display Type Price Quantity & Availability  
Catalog Number Mfr. No. Flow Rate Pressure Material Display Type Price Quantity & Availability  
06-657-005 View Documents Teledyne SSI
LS005SFT1A
5 mL/min 5000 PSI Stainless Steel Pulse Dampener and Pressure Sensor Each for $4,990.00
Add to cart
06-657-006 View Documents Teledyne SSI
LS005PFT3A
5 mL/min 5000 PSI Polyether Ether Ketone (PEEK) Pulse Dampener and Pressure Sensor Each for $4,960.00
Add to cart
06-657-007 View Documents Teledyne SSI
LS010PFT3A
10 mL/min 5000 PSI Polyether Ether Ketone (PEEK) Pulse Dampener and Pressure Sensor Each for $4,990.00
Add to cart
06-657-008 View Documents Teledyne SSI
LS010SFP1A
10 mL/min 6000 PSI Stainless Steel Pulse Dampener Only Each for $4,310.00
Add to cart
06-657-009 View Documents Teledyne SSI
LS010PFP3A
10 mL/min 5000 PSI Polyether Ether Ketone (PEEK) Pulse Dampener Only Each for $4,510.00
Add to cart
06-657-010 View Documents Teledyne SSI
LS040SFT1A
40 mL/min 1600 PSI Stainless Steel Pulse Dampener and Pressure Sensor Each for $5,230.00
Add to cart
06-657-011 View Documents Teledyne SSI
LS040PFT3A
40 mL/min 1600 PSI Polyether Ether Ketone (PEEK) Pulse Dampener and Pressure Sensor Each for $5,230.00
Add to cart
06-657-012 View Documents Teledyne SSI
LS040SFP1A
40 mL/min 1600 PSI Stainless Steel Pulse Dampener Only Each for $4,520.00
Add to cart
06-657-013 View Documents Teledyne SSI
LS040PFP3A
40 mL/min 1600 PSI Polyether Ether Ketone (PEEK) Pulse Dampener Only Each for $4,310.00
Add to cart
Description
Description
  • Self Flush
  • Inlet Tubing:  1/8in. OD Flexible
  • Outlet Tubing:  1/16 in. OD
  • The pumps are ideal for a wide variety of HPLC applications (including Biocompatible separations), Semi-Prep LC, metering & dispensing, and general laboratory use. Standard fluid path materials are Stainless Steel and PEEK, in addition to optional Titanium. Other available features include jacketed heads for temperature controlled processes. With 5 mL/min, 10 mL/min, and 40 mL/min versions available, and pressure capability up to 6,000 psi, these versatile pumps will meet the most demanding applications.
  • Features include automatic pressure compensation, Pulse Dampener, integrated Prime-Purge Valve, interactive keypad control, and complete PC control and status through RS-232 and Micro USB 2.0 ports. The unique LS Class provides the highest performance/cost ratio in a compact package.
Specifications
Specifications
Dispensing, Metering, Semi-Prep, Separation