Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LS1034 (human cecal carcinoma) Whole cell lysates, Non-denatured; Abnova
Click to view available options
Quantity:
200 μg
Description
Western Blotting, Immunoprecipitation
Specifications
Specifications
For Use With (Application) | Immunoprecipitation, Western Blot |
Tissue | Cecum |
Description | Whole cell lysate (non-denatured) |
Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
Preparation Method | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 200 μg |
Storage Buffer | In modified RIPA Lysis Buffer. |
Host Species | Human |
Concentration | 5 mg/mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction