Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSD1, Rabbit anti-Human,Mouse,Rat,Canine,Primate,Zebrafish, DyLight 755, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056475IR

Catalog No. NB003647


Add to cart

Description

Description

LSD1 Polyclonal specifically detects LSD1 in Human, Mouse, Rat, Canine, Primate, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

LSD1
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
amine oxidase (flavin containing) domain 2, AOF2lysine-specific histone demethylase 1, BHC110FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit, Flavin-containing amine oxidase domain-containing protein 2, KIAA0601KDM1, LSD1BRAF35-HDAC complex protein BHC110, lysine (K)-specific demethylase 1, lysine (K)-specific demethylase 1A, lysine-specific histone demethylase 1A
Rabbit
IgG
0.1 mL
DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics
Primary
Human, Mouse, Rat, Canine, Primate, Zebrafish
Western Blot
Western Blot
Affinity Purified, Conjugated
KDM1A
A portion of amino acids 100-150 of human LSD1 was used as the immunogen.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit