Learn More
LSM11 Mouse anti-Human, Clone: OTI2A11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF810041
Catalog No. 50-166-9856
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
LSM11 is a component of the U7 snRNP complex that is involved in the histone 3 ft.-end pre-mRNA processing. Increases U7 snRNA levels but not histone 3 ft.-end pre-mRNA processing activity, when overexpressed. Required for cell cycle progression from G1 to S phases. Binds specifically to the Sm-binding site of U7 snRNA.
Specifications
Specifications
LSM11
Monoclonal
Unconjugated
LSM11
Lyophilized
LSM11
Full length human recombit protein of human LSM11 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2A11
PBS with 8% trehalose and no preservative; pH 7.3
P83369
LSM11, U7 small nuclear RNA associated
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
134353