Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSM12 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548514

Catalog No. PA548514

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse and bovine based on sequence homology.

Specifications

Specifications

LSM12
Polyclonal
PBS with 0.09% sodium azide
Q3MHD2, Q9D0R8
Protein LSM12 homolog, LSM12
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
124801, 268490
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LSM12
Liquid
LSM12
KLH conjugated synthetic peptide between 146-174 amino acids from the C-terminal region of human LSM12.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit