Learn More
Abnova™ LSP1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00004046T01
Catalog No. 89-951-219
Add to cart
Description
Description
Specifications
Specifications
293T
BC001785.1
pCMV-LSP1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LSP1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 37.4kDa
Transient overexpression cell lysate was tested with anti-LSP1 antibody (Mfr. No. H00004046-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human