Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LSP1 Mouse anti-Human, Clone: LSP1/3042, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen LSP1
Clone LSP1/3042
Concentration 0.2 mg/ml
Dilution ELISA, Protein Array
Applications ELISA, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121982
View Documents
Novus Biologicals
NBP30725420UG
20 μg
Each for $138.37
Add to cart
 
NB121981
View Documents
Novus Biologicals
NBP30725410UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

LSP1 Monoclonal antibody specifically detects LSP1 in Human samples. It is validated for ELISA, Protein Array
Specifications

Specifications

LSP1
0.2 mg/ml
ELISA, Peptide Array
Purified
RUO
Human
47 kDa actin binding protein, 47 kDa actin-binding protein, 52 kDa phosphoprotein, F-actin binding and cytoskeleton associated protein, leufactin (leukocyte F-actin binding protein), leukocyte-specific protein 1, lymphocyte-specific protein 1, lymphocyte-specific protein pp52, pp52, WP34Lymphocyte-specific antigen WP34
Recombinant fragment of human LSP1 protein (around aa174-278) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
LSP1/3042
ELISA, Protein Array
Unconjugated
Mouse
Phospho Specific
10 mM PBS with 0.05% BSA
4046
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit