Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LTB4-R1/BLTR Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer: Bioss BS2654R
Description
LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.Specifications
LTB4-R1/BLTR | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
LTB4R | |
Liquid | |
Rabbit | |
IgG | |
100 μL | |
-20°C | |
Primary | |
1241 |
LTB4R | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q15722 | |
Blt; BLT1; BLT-1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; Leukotriene B4 receptor 1; LTB4 receptor; LTB4R; LTB4-R 1; Ltb4r1; LTB4-R1; LTBR1; mBLTR; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7 | |
KLH conjugated synthetic peptide derived from human LTB4-R1. | |
Protein A | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title