Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LTB4R Mouse anti-Human, Clone: 202/7B1, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA528101
Description
LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.Specifications
LTB4R | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q15722 | |
Mouse | |
Protein G | |
RUO | |
1241 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
202/7B1 | |
Unconjugated | |
LTB4R | |
LTB4R | |
HeLa cell transfectants expressing human BLTR | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction