Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTB4R Mouse anti-Human, PE, Clone: 202/7B1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA528102

Catalog No. PIMA528102


Add to cart

Description

Description

LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.
Specifications

Specifications

LTB4R
Monoclonal
1 mg/mL
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Q15722
LTB4R
HeLa cell transfectants expressing human BLTR
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
202/7B1
PE
LTB4R
Lyophilized
Mouse
IgG2a
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
1241
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit